4-Phenylbutyric acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Phenylbutanoic acid
A148817-100G
A148817-10G
A148817-1MG
A148817-5G
A148817-25G
A148817-500G
76689-276EA
53.05
USD
76689-276
76689-278
76689-280
76689-282
77317-560
77549-844
4-Phenylbutyric acid 98%
4-Phenylbutyric acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol |
MDL Number:
MFCD00004403 CAS Number: 1821-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...