Tetraethylene glycol 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, Tetraglycol

A148598-1000G A148598-100G A148598-25G A148598-500G
76689-262EA 30.77 USD
76689-262 76689-264 76689-266 76689-268
Tetraethylene glycol 98%
Tetraethylene glycol

Precaución: for Research Use Only

Formula: C₈H₁₈O₅
MW: 194.23 g/mol
MDL Number: MFCD00002879
CAS Number: 112-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR