Copper(II) sulphate pentahydrate 99%

Supplier: Ambeed
Danger

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

A148190-100G A148190-1000G
77567-060EA 8 USD
77567-060 77600-108
Copper(II) sulphate pentahydrate 99%
Copper(II) sulphate pentahydrate

Precaución: For research use only.

Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
MDL Number: MFCD00149681
CAS Number: 7758-99-8
UN: 3077
ADR: 9 Ⅲ
Merck Index: 13,02682

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR