Bis(diphenylphosphino)acetylene 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Bis(diphenylphosphino)ethyne

A147909-1G A147909-250MG A147909-5G
76689-246EA 77 USD
76689-246 76689-248 77624-336
Bis(diphenylphosphino)acetylene 98%
Bis(diphenylphosphino)acetylene

Precaución: for Research Use Only

Formula: C₂₆H₂₀P₂
MDL Number: MFCD00003045
CAS Number: 5112-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR