trans-4-Propylcyclohexanecarboxylic acid 97%

Supplier: Ambeed
Warning

A147451-100G A147451-10G A147451-25G
76689-238EA 28.65 USD
76689-238 76689-240 76689-242
trans-4-Propylcyclohexanecarboxylic acid 97%
trans-4-Propylcyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₂
MW: 170.24 g/mol
MDL Number: MFCD00059559
CAS Number: 38289-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR