3-(Aminomethyl)-1H-indazole 96%

Supplier: Ambeed
Warning

A147383-100MG A147383-1G A147383-5G
76689-232EA 25.47 USD
76689-232 76689-234 76689-236
3-(Aminomethyl)-1H-indazole 96%
3-(Aminomethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C8H9N3
MW: 147.1772 g/mol
MDL Number: MFCD06738683
CAS Number: 806640-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR