Ethyl methyl carbonate 97%

Supplier: Ambeed
Danger

Synonyms: Carbonic acid ethyl methyl ester

A146927-25G
76689-226EA 6.18 USD
76689-226
Ethyl methyl carbonate 97%
Ethyl methyl carbonate

Precaución: for Research Use Only

Formula: C₄H₈O₃
MW: 104.1045 g/mol
MDL Number: MFCD00191398
CAS Number: 623-53-0
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR