2-Amino-6-(((2-(4-chlorophenyl)thiazol-4-yl)methyl)thio)-4-(4-(2-hydroxyethoxy)phenyl)pyridine-3,5-dicarbonitrile 98+%

Supplier: Ambeed
Warning

Synonyms: Capadenoson

A146488-10MG A146488-1MG A146488-5MG
76689-220EA 135.8 USD
76689-220 76689-222 76689-224
2-Amino-6-(((2-(4-chlorophenyl)thiazol-4-yl)methyl)thio)-4-(4-(2-hydroxyethoxy)phenyl)pyridine-3,5-dicarbonitrile 98+%
2-Amino-6-(((2-(4-chlorophenyl)thiazol-4-yl)methyl)thio)-4-(4-(2-hydroxyethoxy)phenyl)pyridine-3,5-dicarbonitrile

Precaución: for Research Use Only

Formula: C₂₅H₁₈ClN₅O₂S₂
MW: 520.03 g/mol
MDL Number: MFCD09954113
CAS Number: 544417-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR