1-Boc-3-hydroxypiperidine 98%

Supplier: Ambeed
Warning

Synonyms: tert-butyl 3-hydroxypiperidine-1-carboxylate

A146043-1000G A146043-100G A146043-10G A146043-1G A146043-25G A146043-500G A146043-5G
76689-192EA 131 USD
76689-192 76689-194 76689-196 76689-198 76689-200 76689-202 76689-204
1-Boc-3-hydroxypiperidine 98%
1-Boc-3-hydroxypiperidine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
MDL Number: MFCD02093938
CAS Number: 85275-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR