4-Propylbenzaldehyde 97%

Supplier: Ambeed
Warning

A146031-10G A146031-1G A146031-25G A146031-5G
76689-184EA 264.17 USD
76689-184 76689-186 76689-188 76689-190
4-Propylbenzaldehyde 97%
4-Propylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00041870
CAS Number: 28785-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR