(S)-5-(Benzyloxy)-2-(((benzyloxy)carbonyl)amino)-5-oxopentanoic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Benzyloxycarbonyl-L-glutamic acid 5-benzyl ester, Z-Glu(OBzl)-OH, 5-Benzyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic Acid 5-Benzyl Ester
A145641-100G
A145641-10G
A145641-1G
A145641-25G
A145641-5G
76689-164EA
82
USD
76689-164
76689-166
76689-168
76689-170
76689-172
(S)-5-(Benzyloxy)-2-(((benzyloxy)carbonyl)amino)-5-oxopentanoic acid 95%
(S)-5-(Benzyloxy)-2-(((benzyloxy)carbonyl)amino)-5-oxopentanoic acid
Precaución: for Research Use Only
Formula:
C₂₀H₂₁NO₆ |
MDL Number:
MFCD00065697 CAS Number: 5680-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...