Methyl-3-(4-hydroxyphenyl)propionate 98%
Supplier: Ambeed
|
Warning
|
A145502-100G
A145502-10G
A145502-1G
A145502-500G
A145502-5G
A145502-25G
A145502-1000G
76689-148EA
69
USD
76689-148
76689-150
76689-152
76689-154
76689-156
77316-934
77624-206
Methyl-3-(4-hydroxyphenyl)propionate 98%
Methyl-3-(4-hydroxyphenyl)propionate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₃ |
MDL Number:
MFCD00071577 CAS Number: 5597-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...