2,3,5,6-Tetramethylbenzenesulphonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2,3,5,6-Tetramethylbenzene-1-sulphonyl chloride

A144755-5G A144755-25G
76689-134EA 25 USD
76689-134 77879-022
2,3,5,6-Tetramethylbenzenesulphonyl chloride 98%
2,3,5,6-Tetramethylbenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClO₂S
MDL Number: MFCD00014723
CAS Number: 60706-63-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR