7-Bromo-1,1,2-trimethyl-1H-benzo[e]indole 97%

Supplier: Ambeed
Warning

A144013-100MG A144013-1G A144013-250MG A144013-5G
76689-098EA 64 USD
76689-098 76689-100 76689-102 76689-104
7-Bromo-1,1,2-trimethyl-1H-benzo[e]indole 97%
7-Bromo-1,1,2-trimethyl-1H-benzo[e]indole

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrN
MDL Number: MFCD09056715
CAS Number: 879713-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR