trans-2-(Boc-amino)cyclohexanecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: trans-2-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid

A143036-1G A143036-250MG A143036-5G
76689-080EA 84 USD
76689-080 76689-082 77876-982
trans-2-(Boc-amino)cyclohexanecarboxylic acid 98%
trans-2-(Boc-amino)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MDL Number: MFCD02094138
CAS Number: 209128-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR