2,3-Difluoro-4-methoxyaniline 98%

Supplier: Ambeed
Danger

A142581-250MG A142581-5G A142581-10G
76689-040EA 29 USD
76689-040 77624-190 77876-981
2,3-Difluoro-4-methoxyaniline 98%
2,3-Difluoro-4-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇F₂NO
MDL Number: MFCD04115986
CAS Number: 155020-51-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR