6-(p-Tolyl)pyridin-3-amine 98%

Supplier: Ambeed
Warning

A141715-1G A141715-250MG
76689-032EA 97.61 USD
76689-032 76689-034
6-(p-Tolyl)pyridin-3-amine 98%
6-(p-Tolyl)pyridin-3-amine

Precaución: for Research Use Only

Formula: C12H12N2
MW: 184.24 g/mol
MDL Number: MFCD05864839
CAS Number: 170850-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR