Hematoxylin 99%
Supplier: Ambeed
|
Warning
|
A140081-100MG
A140081-50MG
A140081-5G
A140081-1G
A140081-100G
76688-992EA
6
USD
76688-992
76688-994
76688-996
77600-032
77876-970
Hematoxylin 99%
Hematoxylin
Precaución: for Research Use Only
Formula:
C₁₆H₁₄O₆ |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...