6,7-Dichloro-5,10-dihydroimidazo[2,1-b]quinazolin-2(3H)-one hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Anagrelide HCl, Agrylin hydrochloride, 6,7-Dichloro-1,5-dihydroimidazo[2,1-b]quinazolin-2(3H)-one monohydrochloride, BL4162A, Anagrelide hydrochloride, Xagrid hydrochloride

A139945-10MG A139945-1MG A139945-50MG
76688-986EA 47.75 USD
76688-986 76688-988 76688-990
6,7-Dichloro-5,10-dihydroimidazo[2,1-b]quinazolin-2(3H)-one hydrochloride 98%
6,7-Dichloro-5,10-dihydroimidazo[2,1-b]quinazolin-2(3H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₃N₃O
MW: 292.55 g/mol
MDL Number: MFCD01720337
CAS Number: 58579-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR