3-Amino-3-(4-fluorophenyl)propionic acid 97%

Supplier: Ambeed
Warning

A139557-100G A139557-1G A139557-250MG A139557-25G A139557-5G
76688-974EA 342 USD
76688-974 76688-976 76688-978 76688-980 76688-982
3-Amino-3-(4-fluorophenyl)propionic acid 97%
3-Amino-3-(4-fluorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
MDL Number: MFCD00181811
CAS Number: 325-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR