3,3-Pentamethylene glutarimide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Gabapentin Impurity 3
A139387-100G
A139387-1G
A139387-25G
A139387-5G
76688-966EA
134.74
USD
76688-966
76688-968
76688-970
76688-972
3,3-Pentamethylene glutarimide 97%
3,3-Pentamethylene glutarimide
Precaución: for Research Use Only
Formula:
C₁₀H₁₅NO₂ MW: 181.235 g/mol |
MDL Number:
MFCD00023872 CAS Number: 1130-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...