Bithionol 98%

Supplier: Ambeed
Warning

Synonyms: Bithionol, Bis(3,5-dichloro-2-hydroxyphenyl) sulphide, 2,2'-Thiobis(4,6-dichlorophenol), 6,6'-Thiobis(2,4-dichlorophenol)

A138799-1G A138799-1MG A138799-100G A138799-250MG A138799-25G A138799-5G
76688-932EA 11.67 USD
76688-932 76688-934 77624-160 77624-162 77624-164 77624-166
Bithionol 98%
Bithionol

Precaución: for Research Use Only

Formula: C₁₂H₆Cl₄O₂S
MW: 356.0518 g/mol
MDL Number: MFCD00055727
CAS Number: 97-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR