Benzo[b]naphtho[1,2-d]thiophene 97%

Supplier: Ambeed
Warning

A138645-100MG A138645-1G A138645-250MG A138645-25G A138645-5G
76688-914EA 5 USD
76688-914 76688-916 76688-918 76688-920 77624-158
Benzo[b]naphtho[1,2-d]thiophene 97%
Benzo[b]naphtho[1,2-d]thiophene

Precaución: for Research Use Only

Formula: C₁₆H₁₀S
MDL Number: MFCD00215942
CAS Number: 205-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR