tert-Butyl ((1R,3S)-3-hydroxycyclopentyl)carbamate 97%

Supplier: Ambeed
Warning

A138207-1G A138207-250MG A138207-100MG
76688-908EA 98 USD
76688-908 76688-910 77624-152
tert-Butyl ((1R,3S)-3-hydroxycyclopentyl)carbamate 97%
tert-Butyl ((1R,3S)-3-hydroxycyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD18791196
CAS Number: 225641-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR