(±)-δ-Dodecalactone 98%

Supplier: Ambeed
Warning

A138107-100G A138107-25G
76688-902EA 68 USD
76688-902 76688-904 76688-906
(±)-δ-Dodecalactone 98%
(±)-δ-Dodecalactone

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₂
MDL Number: MFCD00006651
CAS Number: 713-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR