tert-Butyl methyl(pyrrolidin-3-yl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: 3-(N-Boc-N-methylamino)pyrrolidine, 3-(N-tert-Butoxycarbonyl-N-methylamino)pyrrolidine

A138042-100MG A138042-1G A138042-250MG A138042-5G
76688-890EA 24 USD
76688-890 76688-892 76688-894 77600-008
tert-Butyl methyl(pyrrolidin-3-yl)carbamate 98%
tert-Butyl methyl(pyrrolidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD00191665
CAS Number: 172478-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR