Boc-7-hydroxy-D-Tic-OH 95%

Supplier: Ambeed
Warning

Synonyms: Boc-D-7-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (R)-2-(tert-Butoxycarbonyl)-7-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

A137927-100MG A137927-5G A137927-1G
76688-886EA 77 USD
76688-886 76688-888 77876-963
Boc-7-hydroxy-D-Tic-OH 95%
Boc-7-hydroxy-D-Tic-OH

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₅
MW: 293.319 g/mol
MDL Number: MFCD00792389
CAS Number: 214630-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR