Bezafibrate 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(2-(4-Chlorobenzamido)ethyl)phenoxy)-2-methylpropanoic acid, 2-[4-[2-(4-Chlorobenzamido)ethyl]phenoxy]isobutyric Acid

A137782-100MG A137782-10G A137782-1G A137782-5G A137782-100G A137782-500G
76688-876EA 5.31 USD
76688-876 76688-878 76688-880 76688-882 77600-004 77624-146
Bezafibrate 98%
Bezafibrate

Precaución: for Research Use Only

Formula: C₁₉H₂₀ClNO₄
MW: 361.82 g/mol
MDL Number: MFCD00078970
CAS Number: 41859-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR