D-β-Homoproline hydrochloride 95%

Supplier: Ambeed
Warning

A137721-100MG A137721-1G A137721-250MG A137721-5G
76688-866EA 16 USD
76688-866 76688-868 76688-870 76688-872
D-β-Homoproline hydrochloride 95%
D-beta-Homoproline hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂·HCl
MDL Number: MFCD06202401
CAS Number: 439918-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR