(R)-(((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphonic acid hydrate 97%

Supplier: Ambeed
Warning

Synonyms: Tenofovir hydrate

A137641-100MG A137641-10MG A137641-1MG A137641-250MG A137641-50MG A137641-5MG
76688-852EA 30.77 USD
76688-852 76688-854 76688-856 76688-858 76688-860 76688-862
(R)-(((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphonic acid hydrate 97%
(R)-(((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphonic acid hydrate

Precaución: for Research Use Only

Formula: C₉H₁₆N₅O₅P
MW: 305.23 g/mol
MDL Number: MFCD11519982
CAS Number: 206184-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR