Ammonium ((1S,3R,4R,7S)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulfonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(-)-3-Bromocamphor-8-sulphonic acid Ammonium salt
A136149-1G
A136149-250MG
76688-818EA
20
USD
76688-818
76688-820
Ammonium ((1S,3R,4R,7S)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulfonate 98%
Ammonium ((1S,3R,4R,7S)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulfonate
Precaución: for Research Use Only
Formula:
C₁₀H₁₈BrNO₄S MW: 328.22 g/mol |
MDL Number:
MFCD00151070 CAS Number: 55870-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...