1-Bromo-4-butylbenzene 98%

Supplier: Ambeed
Warning

A136044-100G A136044-500G A136044-25G A136044-10G
76688-814EA 18 USD
76688-814 76688-816 77298-840 77624-138
1-Bromo-4-butylbenzene 98%
1-Bromo-4-butylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃Br
MDL Number: MFCD00040934
CAS Number: 41492-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR