trans-Cinnamoyl chloride 95%

Supplier: Ambeed
Danger

A135773-25G A135773-5G
76688-800EA 37 USD
76688-800 76688-802
trans-Cinnamoyl chloride 95%
trans-Cinnamoyl chloride

Precaución: for Research Use Only

Formula: C₉H₇ClO
MDL Number: MFCD00000732
CAS Number: 17082-09-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR