tert-Butyl-7-benzyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate 95%

Supplier: Ambeed
Warning

A135716-1G A135716-250MG
76688-796EA 67.9 USD
76688-796 76688-798
tert-Butyl-7-benzyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate 95%
tert-Butyl-7-benzyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₉H₂₈N₂O₂
MW: 316.45 g/mol
MDL Number: MFCD19443918
CAS Number: 227940-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR