7-Bromo-2-methylquinazolin-4(3H)-one 95%

Supplier: Ambeed
Warning

A135590-100MG A135590-1G A135590-250MG A135590-5G
76688-788EA 9 USD
76688-788 76688-790 76688-792 76688-794
7-Bromo-2-methylquinazolin-4(3H)-one 95%
7-Bromo-2-methylquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O
MDL Number: MFCD11043862
CAS Number: 403850-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR