Copper(II) gluconate 98%

Supplier: Ambeed
Warning

A135321-1000G A135321-100G A135321-25G A135321-500G
76688-778EA 83 USD
76688-778 76688-780 76688-782 76688-784
Copper(II) gluconate 98%
Copper(II) gluconate

Precaución: for Research Use Only

Formula: C₁₂H₂₂CuO₁₄
MDL Number: MFCD00075297
CAS Number: 527-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR