3,5-Bis(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A134880-100G A134880-10G A134880-1G A134880-25G A134880-500G
76688-764EA 67.9 USD
76688-764 76688-766 76688-768 76688-770 76688-772
3,5-Bis(trifluoromethyl)benzonitrile 98%
3,5-Bis(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₃F₆N
MW: 239.12 g/mol
MDL Number: MFCD00000379
CAS Number: 27126-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR