1,1'-(Methylenebis(2-ethyl-6-methyl-4,1-phenylene))bis(1H-pyrrole-2,5-dione) 98%

Supplier: Ambeed
Danger

A134309-25G A134309-5G
76688-752EA 29.71 USD
76688-752 76688-754
1,1'-(Methylenebis(2-ethyl-6-methyl-4,1-phenylene))bis(1H-pyrrole-2,5-dione) 98%
1,1'-(Methylenebis(2-ethyl-6-methyl-4,1-phenylene))bis(1H-pyrrole-2,5-dione)

Precaución: for Research Use Only

Formula: C₂₇H₂₆N₂O₄
MW: 442.51 g/mol
MDL Number: MFCD20926792
CAS Number: 105391-33-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR