(R)-tert-Butyl-5-azaspiro[2.4]heptan-7-ylcarbamate 95%

Supplier: Ambeed
Warning

Synonyms: (R)-tert-Butyl 5-azaspiro[2.4]heptan-7-ylcarbamate

A133893-100MG A133893-1G A133893-250MG
76688-738EA 70.02 USD
76688-738 76688-740 76688-742
(R)-tert-Butyl-5-azaspiro[2.4]heptan-7-ylcarbamate 95%
(R)-tert-Butyl-5-azaspiro[2.4]heptan-7-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
MDL Number: MFCD11519122
CAS Number: 127199-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR