Calcium trifluoromethanesulphonate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Calcium trifluoromethanesulphonate, Calcium triflate
A133743-10G
A133743-25G
A133743-5G
76688-734EA
18
USD
76688-734
76688-736
77624-124
Calcium trifluoromethanesulphonate 98%
Calcium trifluoromethanesulphonate
Precaución: for Research Use Only
Formula:
C₂CaF₆O₆S₂ MW: 338.22 |
MDL Number:
MFCD00143911 CAS Number: 55120-75-7 UN: 3260 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...