cis-2,6-Dimethylmorpholine 97%

Supplier: Ambeed
Danger

A133625-1G A133625-25G A133625-5G A133625-10G A133625-100G
76688-724EA 5.31 USD
76688-724 76688-726 76688-728 77566-604 77624-122
cis-2,6-Dimethylmorpholine 97%
cis-2,6-Dimethylmorpholine

Precaución: For research use only.

Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD00078428
CAS Number: 6485-55-8
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR