β-L-Uridine 98%

Supplier: Ambeed
Warning

Synonyms: 1-((2S,3S,4R,5S)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, L-Uridine

A133109-100MG A133109-1G A133109-250MG A133109-5G
76688-716EA 33.95 USD
76688-716 76688-718 76688-720 76688-722
β-L-Uridine 98%
β-L-Uridine

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₆
MW: 244.2 g/mol
MDL Number: MFCD02683606
CAS Number: 26287-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR