7-Chloroindole-3-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 7-Chloro-1H-indole-3-carbaldehyde

A131911-100MG A131911-1G A131911-250MG
76688-660EA 22.28 USD
76688-660 76688-662 76688-664
7-Chloroindole-3-carboxaldehyde 98%
7-Chloroindole-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MW: 179.61 g/mol
MDL Number: MFCD06657153
CAS Number: 1008-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR