2-(Trifluoromethyl)-DL-phenylglycine 98%

Supplier: Ambeed
Warning

Synonyms: H-DL-Phg(2-CF₃)-OH, 2-[Amino(carboxy)methyl]benzotrifluoride, 2-Amino-2-(2-(trifluoromethyl)phenyl)acetic acid, Amino[2-(trifluoromethyl)phenyl]acetic acid

A131275-1G A131275-250MG
76688-630EA 41 USD
76688-630 76688-632
2-(Trifluoromethyl)-DL-phenylglycine 98%
2-(Trifluoromethyl)-DL-phenylglycine

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MW: 219.161 g/mol
MDL Number: MFCD02662403
CAS Number: 240490-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR