Benzeneruthenium(II) chloride dimer 97%

Supplier: Ambeed
Warning

A131225-100MG A131225-1G A131225-250MG A131225-5G A131225-25G
76688-620EA 6 USD
76688-620 76688-622 76688-624 76688-626 76688-628 77624-110
Benzeneruthenium(II) chloride dimer 97%
Benzeneruthenium(II) chloride dimer

Precaución: for Research Use Only

Formula: C₁₂H₁₂Cl₄Ru₂
MDL Number: MFCD00064686
CAS Number: 37366-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR