Sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
β-Glycerophosphate disodium salt pentahydrate
A130983-100G
A130983-10G
A130983-1G
A130983-25G
A130983-5G
A130983-1MG
76688-610EA
120.95
USD
76688-610
76688-612
76688-614
76688-616
76688-618
77624-108
Sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate 95%
Sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate
Precaución: for Research Use Only
Formula:
C₃H₁₇Na₂O₁₁P MW: 306.11 g/mol |
MDL Number:
MFCD00149083 CAS Number: 13408-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...