Naphthalen-2-ylbutanoate 98%

Supplier: Ambeed
Warning

Synonyms: Butyric acid-beta-naphthyl ester, Naphthalen-2-yl butyrate

A130308-1G A130308-250MG
76688-602EA 190.97 USD
76688-602 76688-604
Naphthalen-2-ylbutanoate 98%
Naphthalen-2-ylbutanoate

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.264 g/mol
MDL Number: MFCD00046464
CAS Number: 5856-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR