4-Acetyl-2-fluorobenzonitrile 98%

Supplier: Ambeed
Warning

A128681-100MG A128681-250MG A128681-1G A128681-5G
76688-558EA 17 USD
76688-558 76688-560 77624-094 77876-927
4-Acetyl-2-fluorobenzonitrile 98%
4-Acetyl-2-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₉H₆FNO
MDL Number: MFCD10575008
CAS Number: 214760-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR