Benzyl-4-nitrophenyl carbonate 97%

Supplier: Ambeed
Warning

Synonyms: Benzyl (4-nitrophenyl) carbonate, Carbonic Acid Benzyl 4-Nitrophenyl Ester

A128425-100G A128425-1G A128425-25G A128425-5G
76688-546EA 556.98 USD
76688-546 76688-548 76688-550 76688-552
Benzyl-4-nitrophenyl carbonate 97%
Benzyl-4-nitrophenyl carbonate

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₅
MW: 273.24 g/mol
MDL Number: MFCD00007323
CAS Number: 13795-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR