(S)-(-)-3-Chloro-1-phenyl-1-propanol 95%

Supplier: Ambeed
Warning

A128157-10G A128157-250MG A128157-25G A128157-5G A128157-100G
76688-530EA 24 USD
76688-530 76688-532 76688-534 76688-536 77876-922
(S)-(-)-3-Chloro-1-phenyl-1-propanol 95%
(S)-(-)-3-Chloro-1-phenyl-1-propanol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO
MDL Number: MFCD00013309
CAS Number: 100306-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR